Assay Details
Gene Symbol: cyp26c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029951.2 | NP_001025122.2 | 6 - 7 | 1478 | 62 | |
GenBank mRNA | AY904031.1 | - | 5 - 6 | 1281 | 62 | |
AY920470.1 | - | 5 - 6 | 1303 | 62 | ||
BC129131.1 | - | 6 - 7 | 1478 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
