Assay Details
| Gene Symbol: fynb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029969.1 | NP_001025140.1 | 7 - 8 | 919 | 87 | |
| XM_005160302.4 | XP_005160359.1 | 7 - 8 | 966 | 87 | ||
| XM_021468512.1 | XP_021324187.1 | 6 - 7 | 908 | 87 | ||
| GenBank mRNA | BC098534.1 | - | 7 - 8 | 919 | 87 | |
More Information
Set Membership: |
|

fynb
Genomic Map