Assay Details
| Gene Symbol: fynb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029969.1 | NP_001025140.1 | 8 - 9 | 1086 | 71 | |
| XM_005160302.4 | XP_005160359.1 | 8 - 9 | 1133 | 71 | ||
| XM_021468512.1 | XP_021324187.1 | 7 - 8 | 1075 | 71 | ||
| GenBank mRNA | BC098534.1 | - | 8 - 9 | 1086 | 71 | |
More Information
Set Membership: |
|

fynb
Genomic Map