Assay Details
| Gene Symbol: fynb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029969.1 | NP_001025140.1 | 11 - 12 | 1493 | 73 | |
| XM_005160302.4 | XP_005160359.1 | 11 - 12 | 1540 | 73 | ||
| XM_009294487.3 | XP_009292762.1 | 10 - 11 | 1375 | 73 | ||
| XM_021468512.1 | XP_021324187.1 | 10 - 11 | 1482 | 73 | ||
| GenBank mRNA | BC098534.1 | - | 11 - 12 | 1493 | 73 | |
More Information
Set Membership: |
|

fynb
Genomic Map