Assay Details
| Gene Symbol: golt1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031671.2 | NP_001026841.1 | 2 - 3 | 391 | 144 | |
| GenBank mRNA | BC097251.1 | - | 2 - 3 | 191 | 144 | |
More Information
Set Membership: |
|

golt1bb
Genomic Map