Assay Details
| Gene Symbol: igsf21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034184.2 | NP_001029356.2 | 7 - 8 | 1372 | 60 | |
| GenBank mRNA | BC100052.1 | - | 7 - 8 | 1372 | 60 | |
More Information
Set Membership: |
|

igsf21a
Genomic Map