Assay Details
| Gene Symbol: pfkfb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037377.1 | NP_001032454.1 | 11 - 12 | 1318 | 70 | |
| GenBank mRNA | BC109443.1 | - | 11 - 12 | 1318 | 70 | |
More Information
Set Membership: |
|

pfkfb4a
Genomic Map