Assay Details
| Gene Symbol: crfb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040353.1 | NP_001035443.1 | 2 - 3 | 288 | 71 | |
| GenBank mRNA | BC098871.1 | - | 2 - 3 | 290 | 71 | |
| BC115200.1 | - | 2 - 3 | 288 | 71 | ||
More Information
Set Membership: |
|

crfb12
Genomic Map