Assay Details
| Gene Symbol: trit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044774.1 | NP_001038239.1 | 8 - 9 | 1002 | 68 | |
| XM_021468060.1 | XP_021323735.1 | 6 - 7 | 782 | 68 | ||
| XM_021468061.1 | XP_021323736.1 | 7 - 8 | 955 | 68 | ||
| XM_021468063.1 | XP_021323738.1 | 7 - 8 | 812 | 68 | ||
| XM_021468064.1 | XP_021323739.1 | 5 - 6 | 547 | 68 | ||
| GenBank mRNA | BC091976.1 | - | 8 - 9 | 973 | 68 | |
More Information
Set Membership: |
|

trit1
Genomic Map