Assay Details
| Gene Symbol: sacm1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044878.2 | NP_001038343.2 | 12 - 13 | 1021 | 76 | |
| GenBank mRNA | BC139689.1 | - | 12 - 13 | 1011 | 76 | |
More Information
Set Membership: |
|

sacm1la
Genomic Map