Assay Details
| Gene Symbol: sacm1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044878.2 | NP_001038343.2 | 19 - 20 | 1644 | 102 | |
| GenBank mRNA | BC139689.1 | - | 19 - 20 | 1634 | 102 | |
More Information
Set Membership: |
|

sacm1la
Genomic Map