Assay Details
| Gene Symbol: sacm1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044878.2 | NP_001038343.2 | 2 - 3 | 149 | 62 | |
| GenBank mRNA | BC139689.1 | - | 2 - 3 | 139 | 62 | |
More Information
Set Membership: |
|

sacm1la
Genomic Map