Assay Details
| Gene Symbol: sacm1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044878.2 | NP_001038343.2 | 9 - 10 | 781 | 128 | |
| GenBank mRNA | BC139689.1 | - | 9 - 10 | 771 | 128 | |
More Information
Set Membership: |
|

sacm1la
Genomic Map