Assay Details
| Gene Symbol: nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044993.1 | NP_001038458.1 | 1 - 2 | 286 | 62 | |
| GenBank mRNA | BC134974.1 | - | 1 - 2 | 283 | 62 | |
More Information
Set Membership: |
|

nfs1
Genomic Map