Assay Details
| Gene Symbol: mrps18c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045190.2 | NP_001038655.2 | 2 - 3 | 182 | 71 | |
| GenBank mRNA | BC134080.1 | - | 2 - 3 | 182 | 71 | |
More Information
Set Membership: |
|

mrps18c
Genomic Map