Assay Details
| Gene Symbol: pcgf5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045235.1 | NP_001038700.1 | 3 - 4 | 506 | 93 | |
| XM_005156115.4 | XP_005156172.1 | 3 - 4 | 652 | 93 | ||
| GenBank mRNA | BC116619.1 | - | 3 - 4 | 506 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005156115.4 transcripts.
Set Membership: |
|

pcgf5b
Genomic Map