Assay Details
| Gene Symbol: tle2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045238.1 | NP_001038703.1 | 13 - 14 | 1562 | 67 | |
| XM_009298918.2 | XP_009297193.1 | 13 - 14 | 1674 | 67 | ||
| GenBank mRNA | BC116504.1 | - | 13 - 14 | 1562 | 67 | |
More Information
Set Membership: |
|

tle2c
Genomic Map