Assay Details
| Gene Symbol: abi2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045297.1 | NP_001038762.1 | 1 - 2 | 127 | 87 | |
| XM_005165772.4 | XP_005165829.1 | 1 - 2 | 217 | 87 | ||
| XM_005165774.4 | XP_005165831.1 | 1 - 2 | 216 | 87 | ||
| XM_005165775.4 | XP_005165832.1 | 1 - 2 | 216 | 87 | ||
| XM_005165776.4 | XP_005165833.1 | 1 - 2 | 214 | 87 | ||
| XM_005165777.4 | XP_005165834.1 | 1 - 2 | 214 | 87 | ||
| XM_005165780.4 | XP_005165837.1 | 1 - 2 | 213 | 87 | ||
| XM_009302192.3 | XP_009300467.1 | 1 - 2 | 218 | 87 | ||
| XM_021476926.1 | XP_021332601.1 | 1 - 2 | 217 | 87 | ||
| XM_021476928.1 | XP_021332603.1 | 1 - 2 | 216 | 87 | ||
| XM_021476929.1 | XP_021332604.1 | 1 - 2 | 214 | 87 | ||
| XM_021476930.1 | XP_021332605.1 | 1 - 2 | 214 | 87 | ||
| XM_021476931.1 | XP_021332606.1 | 1 - 2 | 215 | 87 | ||
| XM_021476932.1 | XP_021332607.1 | 1 - 2 | 213 | 87 | ||
| GenBank mRNA | BC116588.1 | - | 1 - 2 | 127 | 87 | |
More Information
Set Membership: |
|

abi2b
Genomic Map