Assay Details
| Gene Symbol: abi2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045297.1 | NP_001038762.1 | 3 - 4 | 476 | 74 | |
| XM_005165774.4 | XP_005165831.1 | 3 - 4 | 565 | 74 | ||
| XM_005165777.4 | XP_005165834.1 | 3 - 4 | 563 | 74 | ||
| XM_021476928.1 | XP_021332603.1 | 3 - 4 | 565 | 74 | ||
| XM_021476930.1 | XP_021332605.1 | 3 - 4 | 563 | 74 | ||
| XM_021476931.1 | XP_021332606.1 | 3 - 4 | 564 | 74 | ||
| XM_021476932.1 | XP_021332607.1 | 3 - 4 | 562 | 74 | ||
| GenBank mRNA | BC116588.1 | - | 3 - 4 | 476 | 74 | |
More Information
Set Membership: |
|

abi2b
Genomic Map