Assay Details
| Gene Symbol: uraha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045452.2 | NP_001038917.2 | 2 - 3 | 288 | 60 | |
| GenBank mRNA | BC095750.1 | - | 2 - 3 | 205 | 60 | |
| BC122322.1 | - | 2 - 3 | 149 | 60 | ||
| DQ640307.1 | - | 2 - 3 | 205 | 60 | ||
| EH526856.1 | - | 2 - 3 | 382 | 60 | ||
More Information
Set Membership: |
|

uraha
Genomic Map