Assay Details
| Gene Symbol: uraha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045452.2 | NP_001038917.2 | 3 - 4 | 418 | 86 | |
| GenBank mRNA | BC095750.1 | - | 3 - 4 | 335 | 86 | |
| BC122322.1 | - | 3 - 4 | 279 | 86 | ||
| DQ640307.1 | - | 3 - 4 | 335 | 86 | ||
| EH526856.1 | - | 1 - 2 | 252 | 86 | ||
More Information
Set Membership: |
|

uraha
Genomic Map