Assay Details
| Gene Symbol: sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045487.2 | NP_001038952.2 | 10 - 11 | 2582 | 59 | |
| GenBank mRNA | BC141794.1 | - | 10 - 11 | 2582 | 59 | |
| BC163246.1 | - | 10 - 11 | 2331 | 59 | ||
More Information
Set Membership: |
|

sh3rf1
Genomic Map