Assay Details
| Gene Symbol: mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045552.1 | NP_001039017.1 | 2 - 3 | 700 | 65 | |
| XM_017351949.2 | XP_017207438.1 | 3 - 4 | 1476 | 65 | ||
| XM_017351950.2 | XP_017207439.1 | 3 - 4 | 1652 | 65 | ||
| XM_017351951.2 | XP_017207440.1 | 3 - 4 | 1494 | 65 | ||
| GenBank mRNA | BC162744.1 | - | 2 - 3 | 787 | 65 | |
| DQ360074.1 | - | 2 - 3 | 700 | 65 | ||
More Information
Set Membership: |
|

mapk6
Genomic Map