Assay Details
| Gene Symbol: pigs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076614.1 | NP_001070082.1 | 3 - 4 | 614 | 66 | |
| XM_009295629.3 | XP_009293904.1 | 4 - 5 | 419 | 66 | ||
| GenBank mRNA | BC124178.1 | - | 3 - 4 | 614 | 66 | |
More Information
Set Membership: |
|

pigs
Genomic Map