Assay Details
| Gene Symbol: rsph10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076759.2 | NP_001070227.1 | 2 - 3 | 347 | 106 | |
| GenBank mRNA | BC124234.1 | - | 2 - 3 | 349 | 106 | |
More Information
Set Membership: |
|

rsph10b
Genomic Map