Assay Details
| Gene Symbol: atp5fa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077355.1 | NP_001070823.1 | 7 - 8 | 1222 | 65 | |
| GenBank mRNA | AY393840.1 | - | 5 - 6 | 925 | 65 | |
| BC124811.1 | - | 7 - 8 | 1222 | 65 | ||
More Information
Set Membership: |
|

atp5fa1
Genomic Map