Assay Details
| Gene Symbol: ctdspl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077544.1 | NP_001071012.1 | 11 - 12 | 1417 | 103 | |
| GenBank mRNA | BC124794.1 | - | 11 - 12 | 1417 | 103 | |
More Information
Set Membership: |
|

ctdspl2a
Genomic Map