Assay Details
| Gene Symbol: scn4ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077570.1 | NP_001071038.1 | 4 - 5 | 1105 | 78 | |
| GenBank mRNA | BC142764.1 | - | 4 - 5 | 1105 | 78 | |
| BC163743.1 | - | 4 - 5 | 813 | 78 | ||
| BC163745.1 | - | 4 - 5 | 813 | 78 | ||
| DQ471305.1 | - | 4 - 5 | 701 | 78 | ||
| DQ489731.1 | - | 4 - 5 | 1105 | 78 | ||
More Information
Set Membership: |
|

scn4ba
Genomic Map