Assay Details
| Gene Symbol: med17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077574.1 | NP_001071042.1 | 4 - 5 | 675 | 58 | |
| GenBank mRNA | BC124507.1 | - | 4 - 5 | 675 | 58 | |
| BC165631.1 | - | 3 - 4 | 651 | 58 | ||
More Information
Set Membership: |
|

med17
Genomic Map