Assay Details
| Gene Symbol: mgat1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079971.1 | NP_001073440.1 | 2 - 3 | 1252 | 75 | |
| XM_005170042.4 | XP_005170099.1 | 2 - 3 | 1324 | 75 | ||
| XM_005170043.4 | XP_005170100.1 | 3 - 4 | 1046 | 75 | ||
| XM_005170044.3 | XP_005170101.1 | 2 - 3 | 1011 | 75 | ||
| GenBank mRNA | BC125909.1 | - | 2 - 3 | 1252 | 75 | |
More Information
Set Membership: |
|

mgat1b
Genomic Map