Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 21 - 22 | 1364 | 114 | |
| XM_009291337.3 | XP_009289612.1 | 21 - 22 | 1630 | 114 | ||
| XM_017351021.2 | XP_017206510.1 | 21 - 22 | 1631 | 114 | ||
| XM_017351022.2 | XP_017206511.1 | 21 - 22 | 1632 | 114 | ||
| XM_017351023.2 | XP_017206512.1 | 21 - 22 | 1626 | 114 | ||
| XM_017351024.2 | XP_017206513.1 | 21 - 22 | 1632 | 114 | ||
| XM_017351025.2 | XP_017206514.1 | 21 - 22 | 1631 | 114 | ||
| XM_017351026.2 | XP_017206515.1 | 21 - 22 | 1616 | 114 | ||
| XM_017351027.2 | XP_017206516.1 | 21 - 22 | 1631 | 114 | ||
| XM_017351028.2 | XP_017206517.1 | 21 - 22 | 1630 | 114 | ||
| XM_017351029.2 | XP_017206518.1 | 21 - 22 | 1631 | 114 | ||
| XM_017351031.2 | XP_017206520.1 | 21 - 22 | 1632 | 114 | ||
| XM_017351032.2 | XP_017206521.1 | 21 - 22 | 1632 | 114 | ||
| XM_017351033.2 | XP_017206522.1 | 17 - 18 | 1352 | 114 | ||
| XM_017351035.2 | XP_017206524.1 | 21 - 22 | 1632 | 114 | ||
| XM_017351036.2 | XP_017206525.1 | 21 - 22 | 1632 | 114 | ||
| XM_017351037.2 | XP_017206526.1 | 21 - 22 | 1632 | 114 | ||
| XM_021481472.1 | XP_021337147.1 | 21 - 22 | 1631 | 114 | ||
| GenBank mRNA | DQ523562.1 | - | 21 - 22 | 1364 | 114 | |
| MF034397.1 | - | 9 - 10 | 575 | 114 | ||
| MF034400.1 | - | 8 - 9 | 544 | 114 | ||
More Information
Set Membership: |
|

elna
Genomic Map