Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 27 - 28 | 1886 | 90 | |
| XM_009291337.3 | XP_009289612.1 | 27 - 28 | 2152 | 90 | ||
| XM_017351021.2 | XP_017206510.1 | 27 - 28 | 2153 | 90 | ||
| XM_017351022.2 | XP_017206511.1 | 27 - 28 | 2148 | 90 | ||
| XM_017351023.2 | XP_017206512.1 | 27 - 28 | 2148 | 90 | ||
| XM_017351024.2 | XP_017206513.1 | 27 - 28 | 2148 | 90 | ||
| XM_017351025.2 | XP_017206514.1 | 27 - 28 | 2153 | 90 | ||
| XM_017351026.2 | XP_017206515.1 | 27 - 28 | 2138 | 90 | ||
| XM_017351027.2 | XP_017206516.1 | 27 - 28 | 2153 | 90 | ||
| XM_017351028.2 | XP_017206517.1 | 27 - 28 | 2152 | 90 | ||
| XM_017351033.2 | XP_017206522.1 | 23 - 24 | 1874 | 90 | ||
| XM_017351038.2 | XP_017206527.1 | 19 - 20 | 1565 | 90 | ||
| XM_021481472.1 | XP_021337147.1 | 27 - 28 | 2111 | 90 | ||
| XM_021481473.1 | XP_021337148.1 | 21 - 22 | 1736 | 90 | ||
| XM_021481474.1 | XP_021337149.1 | 19 - 20 | 1523 | 90 | ||
| GenBank mRNA | DQ523562.1 | - | 27 - 28 | 90 | ||
| MF034400.1 | - | 14 - 15 | 1066 | 90 | ||
| MF039671.1 | - | 4 - 5 | 350 | 90 | ||
| MF039672.1 | - | 4 - 5 | 350 | 90 | ||
| MF039673.1 | - | 4 - 5 | 350 | 90 | ||
| MF039674.1 | - | 4 - 5 | 350 | 90 | ||
More Information
Set Membership: |
|

elna
Genomic Map