Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 36 - 37 | 2460 | 99 | |
| XM_009291337.3 | XP_009289612.1 | 36 - 37 | 2726 | 99 | ||
| XM_017351021.2 | XP_017206510.1 | 36 - 37 | 2727 | 99 | ||
| XM_017351022.2 | XP_017206511.1 | 36 - 37 | 2722 | 99 | ||
| XM_017351023.2 | XP_017206512.1 | 36 - 37 | 2722 | 99 | ||
| XM_017351024.2 | XP_017206513.1 | 36 - 37 | 2722 | 99 | ||
| XM_017351025.2 | XP_017206514.1 | 36 - 37 | 2727 | 99 | ||
| XM_017351026.2 | XP_017206515.1 | 36 - 37 | 2712 | 99 | ||
| XM_017351027.2 | XP_017206516.1 | 36 - 37 | 2727 | 99 | ||
| XM_017351028.2 | XP_017206517.1 | 36 - 37 | 2726 | 99 | ||
| XM_017351029.2 | XP_017206518.1 | 35 - 36 | 2682 | 99 | ||
| XM_017351031.2 | XP_017206520.1 | 34 - 35 | 2599 | 99 | ||
| XM_017351032.2 | XP_017206521.1 | 34 - 35 | 2578 | 99 | ||
| XM_017351033.2 | XP_017206522.1 | 32 - 33 | 2448 | 99 | ||
| XM_017351035.2 | XP_017206524.1 | 32 - 33 | 2449 | 99 | ||
| XM_017351036.2 | XP_017206525.1 | 32 - 33 | 2449 | 99 | ||
| XM_017351037.2 | XP_017206526.1 | 32 - 33 | 2449 | 99 | ||
| XM_017351038.2 | XP_017206527.1 | 28 - 29 | 2139 | 99 | ||
| XM_021481472.1 | XP_021337147.1 | 33 - 34 | 2685 | 99 | ||
| XM_021481473.1 | XP_021337148.1 | 30 - 31 | 2310 | 99 | ||
| XM_021481474.1 | XP_021337149.1 | 25 - 26 | 2097 | 99 | ||
| GenBank mRNA | DQ523562.1 | - | 36 - 37 | 2460 | 99 | |
| MF039657.1 | - | 1 - 2 | 48 | 99 | ||
| MF039658.1 | - | 1 - 2 | 48 | 99 | ||
| MF039659.1 | - | 1 - 2 | 48 | 99 | ||
| MF039660.1 | - | 1 - 2 | 48 | 99 | ||
| MF039661.1 | - | 1 - 2 | 48 | 99 | ||
| MF039662.1 | - | 1 - 2 | 48 | 99 | ||
| MF039663.1 | - | 1 - 2 | 48 | 99 | ||
| MF039664.1 | - | 1 - 2 | 48 | 99 | ||
| MF039671.1 | - | 13 - 14 | 924 | 99 | ||
| MF039672.1 | - | 13 - 14 | 924 | 99 | ||
| MF039673.1 | - | 13 - 14 | 924 | 99 | ||
| MF039674.1 | - | 13 - 14 | 924 | 99 | ||
| MF039676.1 | - | 7 - 8 | 426 | 99 | ||
| MF039677.1 | - | 7 - 8 | 426 | 99 | ||
| MF039678.1 | - | 7 - 8 | 426 | 99 | ||
| MF101833.1 | - | 7 - 8 | 426 | 99 | ||
More Information
Set Membership: |
|

elna
Genomic Map