Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 38 - 39 | 2554 | 103 | |
| XM_009291337.3 | XP_009289612.1 | 38 - 39 | 2820 | 103 | ||
| XM_017351021.2 | XP_017206510.1 | 38 - 39 | 2821 | 103 | ||
| XM_017351022.2 | XP_017206511.1 | 38 - 39 | 2816 | 103 | ||
| XM_017351023.2 | XP_017206512.1 | 38 - 39 | 2816 | 103 | ||
| XM_017351024.2 | XP_017206513.1 | 38 - 39 | 2816 | 103 | ||
| XM_017351025.2 | XP_017206514.1 | 38 - 39 | 2821 | 103 | ||
| XM_017351026.2 | XP_017206515.1 | 38 - 39 | 2806 | 103 | ||
| XM_017351027.2 | XP_017206516.1 | 38 - 39 | 2821 | 103 | ||
| XM_017351028.2 | XP_017206517.1 | 38 - 39 | 2820 | 103 | ||
| XM_017351029.2 | XP_017206518.1 | 37 - 38 | 2776 | 103 | ||
| XM_017351031.2 | XP_017206520.1 | 36 - 37 | 2693 | 103 | ||
| XM_017351032.2 | XP_017206521.1 | 36 - 37 | 2672 | 103 | ||
| XM_017351033.2 | XP_017206522.1 | 34 - 35 | 2542 | 103 | ||
| XM_017351035.2 | XP_017206524.1 | 34 - 35 | 2543 | 103 | ||
| XM_017351036.2 | XP_017206525.1 | 34 - 35 | 2543 | 103 | ||
| XM_017351037.2 | XP_017206526.1 | 34 - 35 | 2543 | 103 | ||
| XM_017351038.2 | XP_017206527.1 | 30 - 31 | 2233 | 103 | ||
| XM_021481472.1 | XP_021337147.1 | 35 - 36 | 2779 | 103 | ||
| XM_021481473.1 | XP_021337148.1 | 32 - 33 | 2404 | 103 | ||
| XM_021481474.1 | XP_021337149.1 | 27 - 28 | 2191 | 103 | ||
| GenBank mRNA | DQ523562.1 | - | 38 - 39 | 2554 | 103 | |
| MF039657.1 | - | 3 - 4 | 142 | 103 | ||
| MF039658.1 | - | 3 - 4 | 142 | 103 | ||
| MF039659.1 | - | 3 - 4 | 142 | 103 | ||
| MF039660.1 | - | 3 - 4 | 142 | 103 | ||
| MF039661.1 | - | 3 - 4 | 142 | 103 | ||
| MF039662.1 | - | 3 - 4 | 142 | 103 | ||
| MF039663.1 | - | 3 - 4 | 142 | 103 | ||
| MF039664.1 | - | 3 - 4 | 142 | 103 | ||
More Information
Set Membership: |
|

elna
Genomic Map