Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 39 - 40 | 2606 | 71 | |
| XM_017351027.2 | XP_017206516.1 | 39 - 40 | 2873 | 71 | ||
| GenBank mRNA | DQ523562.1 | - | 39 - 40 | 2606 | 71 | |
More Information
Set Membership: |
|

elna
Genomic Map