Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 45 - 46 | 3001 | 96 | |
| XM_017351028.2 | XP_017206517.1 | 45 - 46 | 3363 | 96 | ||
| GenBank mRNA | DQ523562.1 | - | 45 - 46 | 3001 | 96 | |
| MF067532.1 | - | 4 - 5 | 310 | 96 | ||
| MF067533.1 | - | 5 - 6 | 329 | 96 | ||
| MF067536.1 | - | 5 - 6 | 329 | 96 | ||
| MF067537.1 | - | 5 - 6 | 329 | 96 | ||
More Information
Set Membership: |
|

elna
Genomic Map