Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 52 - 53 | 3324 | 135 | |
| XM_009291337.3 | XP_009289612.1 | 51 - 52 | 3539 | 135 | ||
| XM_017351021.2 | XP_017206510.1 | 53 - 54 | 3729 | 135 | ||
| XM_017351022.2 | XP_017206511.1 | 53 - 54 | 3724 | 135 | ||
| XM_017351023.2 | XP_017206512.1 | 53 - 54 | 3724 | 135 | ||
| XM_017351024.2 | XP_017206513.1 | 53 - 54 | 3724 | 135 | ||
| XM_017351025.2 | XP_017206514.1 | 53 - 54 | 3717 | 135 | ||
| XM_017351026.2 | XP_017206515.1 | 53 - 54 | 3714 | 135 | ||
| XM_017351027.2 | XP_017206516.1 | 53 - 54 | 3705 | 135 | ||
| XM_017351028.2 | XP_017206517.1 | 52 - 53 | 3686 | 135 | ||
| XM_017351029.2 | XP_017206518.1 | 52 - 53 | 3684 | 135 | ||
| XM_017351031.2 | XP_017206520.1 | 51 - 52 | 3601 | 135 | ||
| XM_017351032.2 | XP_017206521.1 | 51 - 52 | 3580 | 135 | ||
| XM_017351033.2 | XP_017206522.1 | 49 - 50 | 3450 | 135 | ||
| XM_017351035.2 | XP_017206524.1 | 49 - 50 | 3451 | 135 | ||
| XM_017351036.2 | XP_017206525.1 | 49 - 50 | 3451 | 135 | ||
| XM_017351037.2 | XP_017206526.1 | 49 - 50 | 3451 | 135 | ||
| XM_017351038.2 | XP_017206527.1 | 45 - 46 | 3141 | 135 | ||
| XM_021481472.1 | XP_021337147.1 | 50 - 51 | 3687 | 135 | ||
| XM_021481473.1 | XP_021337148.1 | 47 - 48 | 3312 | 135 | ||
| XM_021481474.1 | XP_021337149.1 | 42 - 43 | 3099 | 135 | ||
| GenBank mRNA | DQ523562.1 | - | 52 - 53 | 3324 | 135 | |
| MF067532.1 | - | 11 - 12 | 633 | 135 | ||
| MF067533.1 | - | 12 - 13 | 652 | 135 | ||
| MF067534.1 | - | 12 - 13 | 664 | 135 | ||
| MF067535.1 | - | 12 - 13 | 657 | 135 | ||
| MF067536.1 | - | 12 - 13 | 652 | 135 | ||
| MF067537.1 | - | 12 - 13 | 652 | 135 | ||
| MF067538.1 | - | 13 - 14 | 694 | 135 | ||
More Information
Set Membership: |
|

elna
Genomic Map