Assay Details
| Gene Symbol: elna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080063.2 | NP_001073532.2 | 11 - 12 | 707 | 112 | |
| XM_017351023.2 | XP_017206512.1 | 11 - 12 | 969 | 112 | ||
| GenBank mRNA | DQ523562.1 | - | 11 - 12 | 707 | 112 | |
| MF034394.1 | - | 2 - 3 | 117 | 112 | ||
More Information
Set Membership: |
|

elna
Genomic Map