Assay Details
| Gene Symbol: vhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080684.1 | NP_001074153.1 | 2 - 3 | 567 | 55 | |
| GenBank mRNA | BC129410.1 | - | 2 - 3 | 567 | 55 | |
More Information
Set Membership: |
|

vhl
Genomic Map