Assay Details
| Gene Symbol: rhot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005166191.4 | XP_005166248.1 | 11 - 12 | 1116 | 91 | |
| XM_021476893.1 | XP_021332568.1 | 10 - 11 | 1017 | 91 | ||
| XM_021476894.1 | XP_021332569.1 | 11 - 12 | 1119 | 91 | ||
| XM_021476895.1 | XP_021332570.1 | 5 - 6 | 448 | 91 | ||
More Information
Set Membership: |
|

rhot1b
Genomic Map