Assay Details
| Gene Symbol: rhot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082826.2 | NP_001076295.2 | 13 - 14 | 1316 | 66 | |
| XM_005166191.4 | XP_005166248.1 | 13 - 14 | 1341 | 66 | ||
| XM_021476893.1 | XP_021332568.1 | 12 - 13 | 1242 | 66 | ||
| XM_021476894.1 | XP_021332569.1 | 13 - 14 | 1344 | 66 | ||
| XM_021476895.1 | XP_021332570.1 | 7 - 8 | 673 | 66 | ||
| GenBank mRNA | BC163039.1 | - | 13 - 14 | 1316 | 66 | |
More Information
Set Membership: |
|

rhot1b
Genomic Map