Assay Details
| Gene Symbol: rhot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082826.2 | NP_001076295.2 | 16 - 17 | 1639 | 92 | |
| XM_005166191.4 | XP_005166248.1 | 16 - 17 | 1664 | 92 | ||
| XM_021476893.1 | XP_021332568.1 | 15 - 16 | 1565 | 92 | ||
| XM_021476894.1 | XP_021332569.1 | 16 - 17 | 1667 | 92 | ||
| XM_021476895.1 | XP_021332570.1 | 10 - 11 | 996 | 92 | ||
| GenBank mRNA | BC163039.1 | - | 16 - 17 | 1639 | 92 | |
More Information
Set Membership: |
|

rhot1b
Genomic Map