Assay Details
| Gene Symbol: rhot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082826.2 | NP_001076295.2 | 19 - 20 | 2091 | 91 | |
| XM_005166191.4 | XP_005166248.1 | 19 - 20 | 2116 | 91 | ||
| XM_021476893.1 | XP_021332568.1 | 18 - 19 | 2017 | 91 | ||
| XM_021476895.1 | XP_021332570.1 | 13 - 14 | 1448 | 91 | ||
| GenBank mRNA | BC163039.1 | - | 19 - 20 | 2091 | 91 | |
More Information
Set Membership: |
|

rhot1b
Genomic Map