Assay Details
| Gene Symbol: zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083540.1 | NP_001077009.1 | 5 - 6 | 350 | 76 | |
| GenBank mRNA | BC077127.1 | - | 5 - 6 | 358 | 76 | |
| BC133904.1 | - | 5 - 6 | 350 | 76 | ||
More Information
Set Membership: |
|

zswim7
Genomic Map