Assay Details
| Gene Symbol: rbpjb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083551.1 | NP_001077020.1 | 5 - 6 | 557 | 87 | |
| GenBank mRNA | AY332618.1 | - | 5 - 6 | 557 | 87 | |
| BC163294.1 | - | 5 - 6 | 534 | 87 | ||
| BC163296.1 | - | 5 - 6 | 534 | 87 | ||
| BC167436.1 | - | 5 - 6 | 562 | 87 | ||
| DQ388902.1 | - | 5 - 6 | 559 | 87 | ||
More Information
Set Membership: |
|

rbpjb
Genomic Map