Assay Details
| Gene Symbol: crfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083868.1 | NP_001077337.1 | 4 - 5 | 564 | 110 | |
| GenBank mRNA | BC163069.1 | - | 4 - 5 | 561 | 110 | |
| BC167449.1 | - | 4 - 5 | 618 | 110 | ||
| EF014954.1 | - | 4 - 5 | 564 | 110 | ||
More Information
Set Membership: |
|

crfb4
Genomic Map