Assay Details
| Gene Symbol: axdnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089377.1 | NP_001082846.1 | 1 - 2 | 228 | 56 | |
| GenBank mRNA | BC139551.1 | - | 1 - 2 | 228 | 56 | |
More Information
Set Membership: |
|

axdnd1
Genomic Map