Assay Details
| Gene Symbol: ogfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089549.2 | NP_001083018.1 | 7 - 8 | 688 | 103 | |
| XM_005172867.4 | XP_005172924.1 | 7 - 8 | 686 | 103 | ||
| XM_009300581.3 | XP_009298856.1 | 7 - 8 | 686 | 103 | ||
| GenBank mRNA | BC134915.1 | - | 7 - 8 | 623 | 103 | |
More Information
Set Membership: |
|

ogfr1
Genomic Map