Assay Details
| Gene Symbol: akap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098179.2 | NP_001091649.2 | 4 - 5 | 1966 | 72 | |
| XM_017350962.2 | XP_017206451.1 | 4 - 5 | 1973 | 72 | ||
| GenBank mRNA | BC139889.1 | - | 5 - 6 | 1625 | 72 | |
More Information
Set Membership: |
|

akap1b
Genomic Map