Assay Details
| Gene Symbol: igfbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098257.2 | NP_001091727.1 | 1 - 2 | 403 | 72 | |
| GenBank mRNA | BC135095.1 | - | 1 - 2 | 403 | 72 | |
| DQ465369.1 | - | 1 - 2 | 397 | 72 | ||
More Information
Set Membership: |
|

igfbp1b
Genomic Map